6-Methoxy-3,4-dihydroisoquinolin-1(2H)-one 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR952422-1G OR952422-250MG
APOSOR952422-1GEA 97.9 EUR
APOSOR952422-1G APOSOR952422-250MG
6-Methoxy-3,4-dihydroisoquinolin-1(2H)-one 95%
6-Methoxy-3,4-dihydroisoquinolin-1(2H)-one
Formule: C₁₀H₁₁NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04114865
CAS nummer: 22246-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR