(1-Chloroethyl)benzene 97%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 1-Phenylethyl chloride
OR9525-25ML OR9525-500ML
APOSOR9525-25MLEA 125 EUR
APOSOR9525-25ML APOSOR9525-500ML
(1-Chloroethyl)benzene 97%
(1-Chloroethyl)benzene
Formule: C₆H₅CHClCH₃
MW: 140.61 g/mol
MDL Nummer: MFCD00018870
CAS nummer: 672-65-1