2-Bromo-4-cyanothiazole 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR962783-1G OR962783-5G
APOSOR962783-1GEA 0 EUR
APOSOR962783-1G APOSOR962783-5G
2-Bromo-4-cyanothiazole 96%
2-Bromo-4-cyanothiazole
Formule: C₄HBrN₂S
Kookpunt: 276 °C
Smeltpunt: 60…70 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD09909650
CAS nummer: 848501-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR