2-Pyrrolidin-1-ylmethylpiperidine 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR964117-1G OR964117-250MG OR964117-5G
APOSOR964117-1GEA 198 EUR
APOSOR964117-1G APOSOR964117-250MG APOSOR964117-5G
2-Pyrrolidin-1-ylmethylpiperidine 95%
2-Pyrrolidin-1-ylmethylpiperidine
Formule: C₁₀H₂₀N₂
MDL Nummer: MFCD03426449
CAS nummer: 100158-63-2