Ethyl-2-cyanobenzoate 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
2-Cyanobenzoic acid ethyl ester, Ethyl 2-cyanobenzoate
OR965457-1G
OR965457-5G
APOSOR965457-1GEA
57
EUR
APOSOR965457-1G
APOSOR965457-5G
Ethyl-2-cyanobenzoate 95%
Ethyl-2-cyanobenzoate
Formule:
C₁₀H₉NO₂ MW: 175.19 g/mol |
MDL Nummer:
MFCD03423595 CAS nummer: 6525-45-7 |