3,4-Dichlorothiophenol
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
OR9736-25G
OR9736-1G
OR9736-5G
APOSOR9736-25GEA
64.3
EUR
APOSOR9736-25G
APOSOR9736-1G
APOSOR9736-5G
3,4-Dichlorothiophenol
3,4-Dichlorothiophenol
Formule:
C₆H₄Cl₂S Kookpunt: 119…120 °C/10mmHg Dichtheid: 1,407 g/cm³ (25 °C) Vlampunt: >113 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00004840 CAS nummer: 5858-17-3 EINECS: 227-485-3 UN: 3334 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...