(R)-(-)-ɑ-(Trifluoromethyl)benzyl alcohol
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
PC0788-1G PC0788-5G
APOSPC0788-1GEA 358 EUR
APOSPC0788-1G APOSPC0788-5G
(R)-(-)-ɑ-(Trifluoromethyl)benzyl alcohol
(R)-(-)-alpha-(Trifluoromethyl)benzyl alcohol
Formule: C₈H₇F₃O
Kookpunt: 98…99 °C/17mmHg
Smeltpunt: 25 °C
Dichtheid: 1,3 g/cm³ (25 °C)
Vlampunt: 87 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00077844
CAS nummer: 10531-50-7
Order Now
Specificaties
Environmentally Preferable