α,α,α-Trifluoro-o-toluidine 97%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
PC1020-1KG PC1020-500G
APOSPC1020-1KGEA 95 EUR
APOSPC1020-1KG APOSPC1020-500G
α,α,α-Trifluoro-o-toluidine 97%
α,α,α-Trifluoro-o-toluidine
Useful intermediate for synthesis of quinoline derivatives and other heterocycles. J. Org. Chem., 1994, 59, 5886
Formule: C₇H₆F₃N
Kookpunt: 65…67 °C/15mmHg
Smeltpunt: –34 °C
Dichtheid: 1,282 g/cm³ (25 °C)
Vlampunt: 65 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007718
CAS nummer: 88-17-5
EINECS: 201-806-7
UN: 2942
ADR: 6.1,III