1-Bromo-2,4-difluoro-5-nitrobenzene 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
5-Bromo-2,4-difluoronitrobenzene
PC10283-25G
PC10283-5G
APOSPC10283-25GEA
0
EUR
APOSPC10283-25G
APOSPC10283-5G
1-Bromo-2,4-difluoro-5-nitrobenzene 98%
1-Bromo-2,4-difluoro-5-nitrobenzene
Formule:
C₆H₂BrF₂NO₂ MW: 237.99 g/mol Kookpunt: 85 °C/0.4mmHg Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00129166 CAS nummer: 345-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...