2-[(3,4,4-Trifluoro-3-butenyl)sulphanyl]nicotinohydrazide
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC10346-1G PC10346-500MG
APOSPC10346-1GEA 408 EUR
APOSPC10346-1G APOSPC10346-500MG
2-[(3,4,4-Trifluoro-3-butenyl)sulphanyl]nicotinohydrazide
2-[(3,4,4-Trifluoro-3-butenyl)sulphanyl]nicotinohydrazide
Formule: C₁₀H₁₀F₃N₃OS
MDL Nummer: MFCD01815167
CAS nummer: 478247-67-5