(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I) 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC10734-1G PC10734-5G
APOSPC10734-1GEA 151 EUR
APOSPC10734-1G APOSPC10734-5G
(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I) 95%
(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I)
Formule: C₃₁H₂₃CuF₃N₂P
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28386111
CAS nummer: 1334890-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR