Potassium(3-chlorophenyl)trifluoroborate
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC11229-1G
PC11229-5G
APOSPC11229-1GEA
58.6
EUR
APOSPC11229-1G
APOSPC11229-5G
Potassium(3-chlorophenyl)trifluoroborate
Potassium(3-chlorophenyl)trifluoroborate
Formule:
C₆H₄BClF₃K Smeltpunt: 76…80 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD04115754 CAS nummer: 411206-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...