Fluorocholine chloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: N-(Fluoromethyl)-2-hydroxy-N,N-dimethylethanaminium chloride

PC1838-1G PC1838-250MG
APOSPC1838-1GEA 0 EUR
APOSPC1838-1G APOSPC1838-250MG
Fluorocholine chloride
Fluorocholine chloride
Formule: C₅H₁₃ClFNO
MW: 157.62 g/mol
Smeltpunt: 184…185 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07995053
CAS nummer: 459424-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR