2-Chloro-6-fluorobenzonitrile 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
PC1863-10G
PC1863-25G
APOSPC1863-10GEA
0
EUR
APOSPC1863-10G
APOSPC1863-25G
2-Chloro-6-fluorobenzonitrile 98%
2-Chloro-6-fluorobenzonitrile
Treatment with nucleophiles e.g alkoxides can selectively displace the fluorine atom: Synth. Commun., 621 (1984)
Formule:
C₇H₃ClFN Kookpunt: 104…105 °C/11mmHg Smeltpunt: 55…60 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001780 CAS nummer: 668-45-1 EINECS: 211-571-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...