Potassium (4-acetylphenyl)trifluoroborate
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC200386-1G
PC200386-5G
APOSPC200386-1GEA
24.6
EUR
APOSPC200386-1G
APOSPC200386-5G
Potassium (4-acetylphenyl)trifluoroborate
Potassium (4-acetylphenyl)trifluoroborate
Formule:
C₈H₇BF₃KO Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD07784376 CAS nummer: 252726-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...