6-(tert-Butyl)-8-fluorophthalazin-1(2H)-one 97+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC201088-100MG PC201088-1G PC201088-250MG
APOSPC201088-100MGEA 122 EUR
APOSPC201088-100MG APOSPC201088-1G APOSPC201088-250MG
6-(tert-Butyl)-8-fluorophthalazin-1(2H)-one 97+%
6-(tert-Butyl)-8-fluorophthalazin-1(2H)-one
Formule: C12H13FN2O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD23099569
CAS nummer: 1242156-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR