3-Bromo-5-fluorobenzaldehyde 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PC2392-25G PC2392-5G
APOSPC2392-25GEA 131 EUR
APOSPC2392-25G APOSPC2392-5G
3-Bromo-5-fluorobenzaldehyde 98%
3-Bromo-5-fluorobenzaldehyde
Formule: C₇H₄BrFO
MW: 203.01 g/mol
Smeltpunt: 41…43 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04116319
CAS nummer: 188813-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR