2,4-Dichloro-5-nitrobenzotrifluoride 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
PC2488-100G
PC2488-25G
APOSPC2488-100GEA
0
EUR
APOSPC2488-100G
APOSPC2488-25G
2,4-Dichloro-5-nitrobenzotrifluoride 98%
2,4-Dichloro-5-nitrobenzotrifluoride
Formule:
C₇H₂Cl₂F₃NO₂ Smeltpunt: 53…57 °C Vlampunt: >110 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007073 CAS nummer: 400-70-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...