6-Chloro-2-(trifluoromethyl)imidazo[1,2-b]pyridazine
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC28302-1G PC28302-250MG
APOSPC28302-1GEA 304 EUR
APOSPC28302-1G APOSPC28302-250MG
6-Chloro-2-(trifluoromethyl)imidazo[1,2-b]pyridazine
6-Chloro-2-(trifluoromethyl)imidazo[1,2-b]pyridazine
Formule: C₇H₃ClF₃N₃
MDL Nummer: MFCD07778347
CAS nummer: 109113-97-5