2-Fluoro-4-nitropyridine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PC2992-1G PC2992-250MG
APOSPC2992-1GEA 0 EUR
APOSPC2992-1G APOSPC2992-250MG
2-Fluoro-4-nitropyridine 95%
2-Fluoro-4-nitropyridine
Formule: C₅H₃FN₂O₂
Smeltpunt: 46 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04112489
CAS nummer: 18614-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR