3-(Trifluoromethoxy)benzeneacetyl chloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

PC300007-50MG PC300007-250MG PC300007-500MG
APOSPC300007-50MGEA 124 EUR
APOSPC300007-50MG APOSPC300007-250MG APOSPC300007-500MG
3-(Trifluoromethoxy)benzeneacetyl chloride
3-(Trifluoromethoxy)benzeneacetyl chloride
Formule: C₉H₆ClF₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD17018703
CAS nummer: 929612-79-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR