3-Chloro-4-ethoxy-5-fluorobenzaldehyde 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC302526-10G PC302526-5G PC302526-1G PC302526-250MG
APOSPC302526-10GEA 0 EUR
APOSPC302526-10G APOSPC302526-5G APOSPC302526-1G APOSPC302526-250MG
3-Chloro-4-ethoxy-5-fluorobenzaldehyde 98%
3-Chloro-4-ethoxy-5-fluorobenzaldehyde
Formule: C₉H₈ClFO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06246181
CAS nummer: 883521-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR