4-(3-Chloro-2-fluorophenoxy)-3-fluoroaniline 97%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 3-Fluoro-4-(3-chloro-2-fluorophenoxy)aniline
PC303106-10G PC303106-5G PC303106-1G
APOSPC303106-10GEA 408 EUR
APOSPC303106-10G APOSPC303106-5G APOSPC303106-1G
4-(3-Chloro-2-fluorophenoxy)-3-fluoroaniline 97%
4-(3-Chloro-2-fluorophenoxy)-3-fluoroaniline