Ethyl difluoro(pyridin-2-yl)acetate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PC32718-1G PC32718-5G
APOSPC32718-1GEA 120 EUR
APOSPC32718-1G APOSPC32718-5G
Ethyl difluoro(pyridin-2-yl)acetate 95%
Ethyl difluoro(pyridin-2-yl)acetate
Formule: C₉H₉F₂NO₂
Kookpunt: 79,5 °C/0.5mmHg
Vlampunt: >100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06858718
CAS nummer: 267876-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR