Ishikawa's reagent 25% in acetonitrile
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1-(Diethylamino)-1,1,2,3,3,3-hexafluoropropane, Hexafluoropropene diethylamine, N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine
PC32753-100G PC32753-25G PC32753-500G
APOSPC32753-100GEA 110 EUR
APOSPC32753-100G APOSPC32753-25G APOSPC32753-500G
Ishikawa's reagent 25% in acetonitrile
Ishikawa's reagent
Formule: C₇H₁₁F₆N
MW: 223.16 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00054683
CAS nummer: 309-88-6
UN: 2924
ADR: 3+8 II