Ethyl-4,4,4-trifluoroacetoacetate 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Ethyl trifluoroacetoacetate, Ethyl 4,4,4-trifluoro-3-oxobutanoate
PC3280-100G
PC3280-25G
PC3280-500G
PC3280-5G
APOSPC3280-100GEA
29.2
EUR
APOSPC3280-100G
APOSPC3280-25G
APOSPC3280-500G
APOSPC3280-5G
Ethyl-4,4,4-trifluoroacetoacetate 95%
Ethyl-4,4,4-trifluoroacetoacetate
Formule:
C₆H₇F₃O₃ MW: 184.11 g/mol Kookpunt: 70 °C/100mmHg Smeltpunt: –39 °C Dichtheid: 1.259 g/cm³ (25 °C) Vlampunt: 38 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000424 CAS nummer: 372-31-6 EINECS: 206-750-7 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...