2-Chloro-6-fluoroquinoline 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC404621-1G PC404621-250MG
APOSPC404621-1GEA 114 EUR
APOSPC404621-1G APOSPC404621-250MG
2-Chloro-6-fluoroquinoline 98%
2-Chloro-6-fluoroquinoline
Formule: C₉H₅ClFN
MDL Nummer: MFCD08236768
CAS nummer: 77119-53-0