4-(4-Fluorophenoxy)benzonitrile
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC410059-1G PC410059-5G
APOSPC410059-1GEA 65.8 EUR
APOSPC410059-1G APOSPC410059-5G
4-(4-Fluorophenoxy)benzonitrile
4-(4-Fluorophenoxy)benzonitrile
Formule: C₁₃H₈FNO
MDL Nummer: MFCD01815090
CAS nummer: 215589-24-5