(2-Fluorophenyl)acetic acid 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
PC4120-100G
PC4120-1KG
PC4120-500G
APOSPC4120-100GEA
32.2
EUR
APOSPC4120-100G
APOSPC4120-1KG
APOSPC4120-500G
(2-Fluorophenyl)acetic acid 98%
(2-Fluorophenyl)acetic acid
Formule:
(C₆H₄F)CH₂COOH Smeltpunt: 61…65 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004315 CAS nummer: 451-82-1 EINECS: 207-196-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...