2-(4-Fluorophenyl)indole 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
PC4160-1G
PC4160-5G
PC4160-25G
APOSPC4160-1GEA
22
EUR
APOSPC4160-1G
APOSPC4160-5G
APOSPC4160-25G
2-(4-Fluorophenyl)indole 97%
2-(4-Fluorophenyl)indole
Formule:
C₁₄H₁₀FN MW: 211.24 g/mol Smeltpunt: 190…192 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00068178 CAS nummer: 782-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...