Potassium benzyltrifluoroborate 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
PC50055-5G
PC50055-10G
APOSPC50055-5GEA
76
EUR
APOSPC50055-5G
APOSPC50055-10G
Potassium benzyltrifluoroborate 97%
Potassium benzyltrifluoroborate
Formule:
C₇H₇BF₃K Smeltpunt: >300 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD03427261 CAS nummer: 329976-73-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...