Manganese(III) fluoride 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

PC5040-10G PC5040-50G
APOSPC5040-10GEA 123 EUR
APOSPC5040-10G APOSPC5040-50G
Manganese(III) fluoride 99%
Manganese(III) fluoride
Formule: F₃Mn
MW: 111.93 g/mol
Dichtheid: 3.54 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016223
CAS nummer: 7783-53-1
EINECS: 232-006-6
UN: 3087
ADR: 5.1+6.1 II
Merck Index: 12,05784

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR