3,3,4,4,5,5,5-Heptafluoro-2-pentanone 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Methyl heptafluoropropylketone

PC5113-1G PC5113-5G
APOSPC5113-1GEA 27.8 EUR
APOSPC5113-1G APOSPC5113-5G
3,3,4,4,5,5,5-Heptafluoro-2-pentanone 97%
3,3,4,4,5,5,5-Heptafluoro-2-pentanone
Formule: C₅H₃F₇O
MW: 212.07 g/mol
Kookpunt: 63…64 °C
Vlampunt: 21–23 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042079
CAS nummer: 355-17-9
UN: 1224
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR