α-Methyl-4-trifluoromethylbenzyl alcohol 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PC5174-5G PC5174-25G
APOSPC5174-5GEA 136 EUR
APOSPC5174-5G APOSPC5174-25G
α-Methyl-4-trifluoromethylbenzyl alcohol 96%
alpha-Methyl-4-trifluoromethylbenzyl alcohol
Formule: C₉H₉F₃O
Kookpunt: 62…63 °C/0,5mmHg
Dichtheid: 1,237 g/cm³ (25 °C)
Vlampunt: 98 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00039618
CAS nummer: 1737-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR