4-Chloro-2,6-difluorobenzonitrile 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC53003-5G PC53003-1G
APOSPC53003-5GEA 104 EUR
APOSPC53003-5G APOSPC53003-1G
4-Chloro-2,6-difluorobenzonitrile 98%
4-Chloro-2,6-difluorobenzonitrile
Formule: C₇H₂ClF₂N
MDL Nummer: MFCD06660237
CAS nummer: 886500-41-0