1,1,1,5,5,6,6,6-Octafluoro-2,4-hexanedione 97%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC5336-1G PC5336-5G
APOSPC5336-1GEA 388 EUR
APOSPC5336-1G APOSPC5336-5G
1,1,1,5,5,6,6,6-Octafluoro-2,4-hexanedione 97%
1,1,1,5,5,6,6,6-Octafluoro-2,4-hexanedione
Formule: C₆H₂F₈O₂
CAS nummer: 20825-07-4