Pentafluorophenylhydrazine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PC5690-25G PC5690-5G PC5690-100G
APOSPC5690-25GEA 190 EUR
APOSPC5690-25G APOSPC5690-5G APOSPC5690-100G
Pentafluorophenylhydrazine 97%
Pentafluorophenylhydrazine
Formule: C₆H₃F₅N₂
Smeltpunt: 74…76 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007574
CAS nummer: 828-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR