1-Iodoperfluorodecane 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Henicosafluoro-10-iododecane, Perfluorodecyl iodide, Perfluoro-1-iododecane

PC5970-1G PC5970-25G PC5970-100G
APOSPC5970-1GEA 22 EUR
APOSPC5970-1G APOSPC5970-25G APOSPC5970-100G
1-Iodoperfluorodecane 98%
1-Iodoperfluorodecane
Formule: C₁₀F₂₁I
Kookpunt: 195…200 °C
Smeltpunt: 63…67 °C
Dichtheid: 1,94 g/cm³ (25 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00001065
CAS nummer: 423-62-1
EINECS: 207-030-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR