Trifluoro(trifluoromethoxy)ethylene 99%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Trifluoromethyl trifluorovinyl ether
PC7778E-100G PC7778E-25G
APOSPC7778E-100GEA 935 EUR
APOSPC7778E-100G APOSPC7778E-25G
Trifluoro(trifluoromethoxy)ethylene 99%
Trifluoro(trifluoromethoxy)ethylene
Improves low temperature performance & chemical resistance when used as a co-polymer to VDF &/or PTFE.
Formule: C₃F₆O
MW: 166.02 g/mol
Kookpunt: –23 °C
Smeltpunt: -125
Dichtheid: 1.43 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00221780
CAS nummer: 1187-93-5
EINECS: 214-703-7
UN: 3153
ADR: 2.1