Trimethyloxonium tetrafluoroborate 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
PC7865-100G PC7865-25G PC7865-5G
APOSPC7865-100GEA 101 EUR
APOSPC7865-100G APOSPC7865-25G APOSPC7865-5G
Trimethyloxonium tetrafluoroborate 95%
Trimethyloxonium tetrafluoroborate
Versatile mild methylating agent
Formule: C₃H₉BF₄O
MW: 147.91 g/mol
Smeltpunt: 200 °C
Vlampunt: >110 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00011798
CAS nummer: 420-37-1
EINECS: 206-994-4
UN: 3261
ADR: 8 II