Tris(pentafluorophenyl)phosphine 99%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Perfluorotriphenylphosphine, Tris(perfluorophenyl)phosphine
PC7905-1G PC7905-5G PC7905-25G
APOSPC7905-1GEA 22 EUR
APOSPC7905-1G APOSPC7905-5G APOSPC7905-25G
Tris(pentafluorophenyl)phosphine 99%
Tris(pentafluorophenyl)phosphine
Formule: C₁₈F₁₅P
MW: 532.15 g/mol
Smeltpunt: 108…110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00079654
CAS nummer: 1259-35-4
EINECS: 215-021-2