N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TSTU, 2-(2,5-Dioxopyrrolidin-1-yl)-1,1,3,3-tetramethylisouronium tetrafluoroborate
PC8350-100G PC8350-25G PC8350-5G
APOSPC8350-100GEA 70.1 EUR
APOSPC8350-100G APOSPC8350-25G APOSPC8350-5G
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate 98%
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate
Formule: C₉H₁₆BF₄N₃O₃
MW: 301.05 g/mol
Smeltpunt: 195…201 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00077875
CAS nummer: 105832-38-0