3-Fluorothiophene-2-carboxylic acid
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
3-Fluoro-2-thiophenecarboxylic acid
PC8915-10G
PC8915-1G
PC8915-5G
APOSPC8915-10GEA
269
EUR
APOSPC8915-10G
APOSPC8915-1G
APOSPC8915-5G
3-Fluorothiophene-2-carboxylic acid
3-Fluorothiophene-2-carboxylic acid
Formule:
C5H3FO2S Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD03426911 CAS nummer: 32431-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...