2,4-Difluorobenzoylacetonitrile 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
PC9043-1G
PC9043-5G
APOSPC9043-1GEA
87.6
EUR
APOSPC9043-1G
APOSPC9043-5G
2,4-Difluorobenzoylacetonitrile 97%
2,4-Difluorobenzoylacetonitrile
Formule:
C₉H₅F₂NO Smeltpunt: 107…110 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD02260804 CAS nummer: 71682-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...