2-(5-Fluoropyridin-3-yl)acetic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC904503-1G PC904503-250MG
APOSPC904503-1GEA 372 EUR
APOSPC904503-1G APOSPC904503-250MG
2-(5-Fluoropyridin-3-yl)acetic acid 98%
2-(5-Fluoropyridin-3-yl)acetic acid
Formule: C₇H₆FNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09924977
CAS nummer: 38129-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR