4-(Trifluoromethyl)cyclohexanemethanol (cis- and trans- mixture) 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: [4-(Trifluoromethyl)cyclohexyl]methanol
PC906474-1G PC906474-5G
APOSPC906474-1GEA 337 EUR
APOSPC906474-1G APOSPC906474-5G
4-(Trifluoromethyl)cyclohexanemethanol (cis- and trans- mixture) 95%
4-(Trifluoromethyl)cyclohexanemethanol (cis- and trans- mixture)
Formule: C₈H₁₃F₃O
MW: 182.19 g/mol
CAS nummer: 883731-58-6