α,α,α,3-Tetrafluoro-p-toluic acid 99%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC9115-100G PC9115-25G PC9115-500G
APOSPC9115-100GEA 113 EUR
APOSPC9115-100G APOSPC9115-25G APOSPC9115-500G
α,α,α,3-Tetrafluoro-p-toluic acid 99%
α,α,α,3-Tetrafluoro-p-toluic acid
Formule: C₈H₄F₄O₂
MW: 208.11 g/mol
MDL Nummer: MFCD00236279
CAS nummer: 115754-21-7