4-(4-Fluorophenoxy)-3-nitrobenzaldehyde 97%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC9350-10G PC9350-1G PC9350-5G
APOSPC9350-10GEA 1230 EUR
APOSPC9350-10G APOSPC9350-1G APOSPC9350-5G
4-(4-Fluorophenoxy)-3-nitrobenzaldehyde 97%
4-(4-Fluorophenoxy)-3-nitrobenzaldehyde
Formule: C₁₃H₈FNO₄
MDL Nummer: MFCD00138746
CAS nummer: 320416-60-2