6-Oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC9476-1G PC9476-500MG PC9476-5G
APOSPC9476-1GEA 173 EUR
APOSPC9476-1G APOSPC9476-500MG APOSPC9476-5G
6-Oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid 97%
6-Oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid
Formule: C₁₄H₁₀F₃NO₃
Smeltpunt: 251…253 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00243721
CAS nummer: 338783-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR