2-Bromo-5-fluoropyridine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PC9922-100G PC9922-25G PC9922-5G PC9922-1G
APOSPC9922-100GEA 134 EUR
APOSPC9922-100G APOSPC9922-25G APOSPC9922-5G APOSPC9922-1G
2-Bromo-5-fluoropyridine 98%
2-Bromo-5-fluoropyridine
Formule: C₅H₃BrFN
Kookpunt: 80…83 °C/44mmHg
Smeltpunt: 26…31 °C
Vlampunt: 73 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00234011
CAS nummer: 41404-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR